missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Density Standards (100 °C), Paragon Scientific™
Paragon ISO 17025 / ISO 17034 certified Density standards are for the verification of instruments used to measure density of materials which are fluid at the desired test temperature within the range of 15°C to 150°C
£70.40
Specifications
Quantity | 60 mL |
---|---|
Certifications/Compliance | Dual certified to ISO 17025 / ISO 17034 under our UKAS accreditation |
Product Code | Brand | Density | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Density | Price | Quantity & Availability | |||||
13420539
|
Paragon Scientific
DEN100-01 |
0.768 g/mL (100°C) |
£70.40
60mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
13430539
|
Paragon Scientific
DEN100-02 |
0.8103 g/mL (100°C) |
£70.40
60mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
13440539
|
Paragon Scientific
DEN100-03 |
0.8541 g/mL (100°C) |
£70.40
60mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- Dual certified to ISO 17025 / ISO 17034 under our UKAS accreditation
- Certified in accordance with primary level ASTM D1480 methodology
- Ensures full compliance to ASTM and IP test method protocols.
- Supplied in tamper evident packaging with a 2 year shelf life.
- Fully traceable to international standards.
- Supplied with MSDS.
- Tamper evident security packaging.
- Manufactured in the United Kingdom
Specifications
60 mL | |
Dual certified to ISO 17025 / ISO 17034 under our UKAS accreditation |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- DEN100-01
Signal Word
- Danger
Hazard Category
- Aspiration toxicity Category 1
Hazard Statement
- H304-May be fatal if swallowed and enters airways.
Precautionary Statement
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P331-Do NOT induce vomiting.
Supplemental information
- MIXTURE LIST-Contains : Kerosine sweetened
Nominal values quoted