Learn More
Density Standards (15 °C), Paragon Scientific™
Paragon ISO 17025 / ISO 17034 certified Density standards are for the verification of instruments used to measure density of materials which are fluid at the desired test temperature within the range of 15°C
Brand:  Paragon Scientific DEN15-12
 View more versions of this product
Product Code. 13420489
  • £70.40 / 60mL
Explore more special offers
Description
Description
  • Dual certified to ISO 17025 / ISO 17034 under our UKAS accreditation
  • Certified in accordance with primary level ASTM D1480 methodology
  • Ensures full compliance to ASTM and IP test method protocols.
  • Supplied in tamper evident packaging with a 2 year shelf life.
  • Fully traceable to international standards.
  • Supplied with MSDS.
  • Tamper evident security packaging.
  • Manufactured in the United Kingdom
Specifications
Specifications
60 mL
1.1280 g/mL (15°C)
Dual certified to ISO 17025 / ISO 17034 under our UKAS accreditation
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Nominal values quoted