Learn More
Density Standards (20 °C), Paragon Scientific™
Paragon ISO 17025 / ISO 17034 certified Density standards are for the verification of instruments used to measure density of materials which are fluid at the desired test temperature within the range of 20°C
£68.00 - £98.90
Product Code | Brand | Density | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Density | Price | Quantity & Availability | |||||
13480489
|
Paragon Scientific
DEN20-01 |
0.6597 g/mL (20°C) |
£68.00
60mL |
Estimated Shipment: 05-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
13470489
|
Paragon Scientific
DEN20-02 |
0.7264 g/mL (20°C) |
£68.00
60mL |
Estimated Shipment: 05-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
13490489
|
Paragon Scientific
DEN20-03 |
0.7788 g/mL (20°C) |
£71.00
60mL |
Estimated Shipment: 05-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
13400499
|
Paragon Scientific
DEN20-04 |
0.8383 g/mL (20°C) |
£81.90
60mL |
Estimated Shipment: 27-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
13420499
|
Paragon Scientific
DEN20-05 |
0.8556 g/mL (20°C) |
£71.55
60mL |
Estimated Shipment: 27-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
13450499
|
Paragon Scientific
DEN20-06 |
0.8708 g/mL (20°C) |
£68.00
60mL |
Estimated Shipment: 05-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
13440499
|
Paragon Scientific
DEN20-07 |
0.9376 g/mL (20°C) |
£74.25
60mL |
Estimated Shipment: 05-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
13430499
|
Paragon Scientific
DEN20-08 |
0.9817 g/mL (20°C) |
£71.00
60mL |
Estimated Shipment: 05-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
13460499
|
Paragon Scientific
DEN20-09 |
1.0257 g/mL (20°C) |
£98.70
60mL |
Estimated Shipment: 05-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
13490499
|
Paragon Scientific
DEN20-10 |
1.0677 g/mL (20°C) |
£68.00
60mL |
Estimated Shipment: 05-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
13480499
|
Paragon Scientific
DEN20-11 |
1.1258 g/mL (20°C) |
£98.90
60mL |
Estimated Shipment: 02-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
13400509
|
Paragon Scientific
DEN20-12 |
1.1917 g/mL (20°C) |
£88.10
60mL |
Estimated Shipment: 12-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
13410509
|
Paragon Scientific
DEN20-13 |
1.2849 g/mL (20°C) |
£86.35
60mL |
Estimated Shipment: 02-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
13420509
|
Paragon Scientific
DEN20-14 |
1.6226 g/mL (20°C) |
£82.00
60mL |
Estimated Shipment: 05-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- Dual certified to ISO 17025 / ISO 17034 under our UKAS accreditation
- Certified in accordance with primary level ASTM D1480 methodology
- Ensures full compliance to ASTM and IP test method protocols.
- Supplied in tamper evident packaging (please refer to standard for shelf life).
- Manufactured in the United Kingdom and available from stock for immediate dispatch
Nominal values quoted