missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Density Standards (25 °C), Paragon Scientific™
Paragon ISO 17025 / ISO 17034 certified Density standards are for the verification of instruments used to measure density of materials which are fluid at the desired test temperature within the range of 25°C
Marque: Paragon Scientific DEN25-02
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
- Dual certified to ISO 17025 / ISO 17034 under our UKAS accreditation
- Certified in accordance with primary level ASTM D1480 methodology
- Ensures full compliance to ASTM and IP test method protocols.
- Supplied in tamper evident packaging with a 2 year shelf life.
- Fully traceable to international standards.
- Supplied with MSDS.
- Tamper evident security packaging.
- Manufactured in the United Kingdom
Spécification
60 mL | |
0.7223 g/mL (25°C) | |
Dual certified to ISO 17025 / ISO 17034 under our UKAS accreditation |
Sécurité et traitement
Identifiant produit
Mention d'avertissement
Catégorie de danger
Mention de danger
Conseils de prudence
Informations supplémentaires
- DEN25-02
Mention d'avertissement
- Danger
Catégorie de danger
- Aspiration toxicity Category 1
- LONG-TERM AQUATIC HAZARD Chronic 1
- Flammable liquid Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Mention de danger
- H225-Highly flammable liquid and vapour.
- H304-May be fatal if swallowed and enters airways.
- H315-Causes skin irritation.
- H336-May cause drowsiness or dizziness.
- H410-Very toxic to aquatic life with long lasting effects.
Conseils de prudence
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P331-Do NOT induce vomiting.
Informations supplémentaires
- MIXTURE LIST-Contains : Kerosine : distillates, hydrotreated light
Nominal values quoted