Learn More
MedChemExpress Deoxycytidine triphosphate
MedChemExpress Deoxycytidine triphosphate (dCTP) is a nucleoside triphosphate that can be used for DNA synthesis. Deoxycytidine triphosphate has many applications, such as real-time PCR, cDNA synthesis, and DNA sequencing.
£110.11 - £228.69
Specifications
CAS | 2056-98-6 |
---|---|
Content And Storage | Powder -20°C 3 years, 4°C 2 years ∣ In solvent -80°C 6 months, -20°C 1 month |
Product Type | Biochemical Assay Reagents |
Shipping Condition | Room temperature |
Synonym | dCTP ∣ 2′-Deoxycytidine-5′-triphosphate |
Specifications
2056-98-6 | |
Biochemical Assay Reagents | |
dCTP ∣ 2′-Deoxycytidine-5′-triphosphate | |
H2O : 50 mg/mL (107.03 mM; Need ultrasonic) |
Powder -20°C 3 years, 4°C 2 years ∣ In solvent -80°C 6 months, -20°C 1 month | |
Room temperature | |
467.16 g/mol |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.