missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Deoxynivalenol, Tocris Bioscience™
Click to view available options
Quantity:
1 mg
Description
Tricothecene mycotoxin and potent protein synthesis inhibitor. Exhibits cytotoxic activity in vivo via the ribotoxic stress response. Induces p38-mediated gene expression and apoptosis in leukocytes; activity results in systemic expression of interleukin-6 (IL-6) and other proinflammatory cytokines. Also induces migration of NF-κB into the nucleus.
Specifications
CAS | 51481-10-8 |
Molecular Formula | C15H20O6 |
MDL Number | MFCD09039270 |
Quantity | 1 mg |
Synonym | deoxynivalenol, vomitoxin, dehydronivalenol, 4-deoxynivalenol, 4-desoxynivalenol, desoxynivalenol, unii-jt37hyp23v, ccris 7801, rd toxin, jt37hyp23v |
InChI Key | LINOMUASTDIRTM-QGRHZQQGSA-N |
SMILES | CC1=C[C@H]2O[C@@H]3[C@H](O)C[C@@](C)([C@]33CO3)[C@@]2(CO)[C@H](O)C1=O |
IUPAC Name | (1'R,2S,2'R,3'S,7'R,9'R,10'R)-3',10'-dihydroxy-2'-(hydroxymethyl)-1',5'-dimethyl-8'-oxaspiro[oxirane-2,12'-tricyclo[7.2.1.0²,⁷]dodecan]-5'-en-4'-one |
Molecular Weight (g/mol) | 296.32 |
PubChem CID | 40024 |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'