missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DEPC-Treated Water, MP Biomedicals™
Click to view available options
Quantity:
5 x 200 mL
Specifications
CAS | 7732-18-5 |
Molecular Formula | H2O |
Synonym | dihydrogen oxide, dihydrogen monoxide |
InChI Key | XLYOFNOQVPJJNP-UHFFFAOYSA-N |
SMILES | O |
IUPAC Name | oxidane |
Molecular Weight (g/mol) | 18.015 |
PubChem CID | 962 |
ChEBI | CHEBI:15377 |
Formula Weight | 18.02 |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'