missing translation for 'onlineSavingsMsg'
Learn More

asecos™ Depot PE with Pull-Down Blind - Version Drum

The Depot PE with Pull-Down Blind is approved for the safe storage of water pollutants and has a high resistance to oils, acids and alkalis.

POA

Specifications

Color Grey
Capacity (English) 64.7 gal.
Capacity (Metric) 245 L
Material Polyethylene
Type Disposal Can
View More Specs

Products 2
Product Code Brand Depth (Metric) Price Quantity & Availability  
Product Code Brand Depth (Metric) Price Quantity & Availability  
17836456
View Documents
asecos™
40060
1050.00 mm
POA
Each
N/A
This item requires a price update prior to being ordered.

Request a Price for this item here.

17836586
View Documents
asecos™
40065
1083.00 mm
POA
Each
N/A
This item requires a price update prior to being ordered.

Request a Price for this item here.

Description

Description

  • With integrated sump pallet
  • For direct placement of a barrel on a Euro pallet or barrel storage on the optional PE half pallet
  • Integrated pocket for access underneath by forklifts and forklift trucks
  • Ready for anchoring to the ground (anchor kit optional)
Specifications

Specifications

Grey
245 L
Disposal Can
64.7 gal.
Polyethylene
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
asecos™ Depot PE with Pull-Down Blind - Version Drum

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.