missing translation for 'onlineSavingsMsg'
Learn More

Ansell™ DermaShield™ 73-721 Series Green Neoprene Gloves

Product Code. 12465080 Shop All Ansell Products
Click to view available options
Size:
6
6.5
7
7.5
8
8.5
9
Unit Size:
200 pairs
This item is not returnable. View return policy

Product Code. 12465080

Brand: Ansell™ 73721075

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Offers the comfort and flexibility of natural rubber gloves, but neoprene composition provides better chemical resistance. Free of latex proteins and accelerators, it minimizes both Type I and Type IV allergies associated with natural rubber.

  • Made from poly-chloroprene
  • For non-sterile controlled environments
  • Free from latex proteins and accelerators
  • Neoprene coating offers chemical resistance
  • Rolled beaded cuff prevents glove from rolling down
  • Bisque fingertip grip
  • Powder-free
TRUSTED_SUSTAINABILITY

Specifications

Back Material Neoprene
Cuff Material Neoprene
Applications Industrial
Color Green
Latex Free Yes
Material Polychloroprene
Product Type Neoprene Gloves
Size 7.5
Thickness (Metric) 0.175 mm
Glove Cuff Color Green
Glove Cuff Style Beaded/Rolled
Quantity 1 Pair
Resistant To Chemical resistance
Series 73-721
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.