Learn More
Thermo Scientific™ Dermatophyte Selective Agar (Taplin)
Description
Selectively isolate and rapidly differentiate dermatophytes and other fungi with Thermo Scientific™ Dermatophyte Selective Agar (Taplin). The medium contains cycloheximide, gentamycin and chlortetracycline which suppress accompanying bacterial flora, as well as partially suppressing the growth of other yeasts and molds. A color change in the medium differentiates dermophytes from other fungi that may be present.
In addition to the selective inhibitors, cycloheximide, gentamycin and chlortetracycline, Dermatophyte Selective Agar as per Taplin contains phenol red pH indicator. A color change from yellow to red indicates the proteolytic activity of most dermatophytes and the formation of basic metabolites. In contrast to typical dermatophytes, many other molds form acidic metabolites which do not change the color of the agar. It is, therefore, possible to rapidly differentiate between dermatophytes and other fungi.
- Selective: accompanying bacterial flora are suppressed
- Clear results: color change aids rapid differentiation of dermatophytes
- Ready-to-use: convenience of prepared media
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
Specifications
Specifications
| Content And Storage | 6°C to 12°C |
| Description | Dermatophyte Selective Agar (Taplin) |
| Format | 90mm Monoplate |
| Product Type | Agar |
| Fill Weight | 17.5g ± 5% |
| Packaging Type | Wrapped in single cellulose-based film wrap |
| Quantity | 10/Pk. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.