missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Honeywell KCL™ Dermatril™ P 743 Gloves
Click to view available options
Size:
6
7
8
9
10
11
Description
- Disposable protective glove according to EN 374
- The chemical as well as the mechanical resistance are increased compared to the Dermatril™ 740
- Can be used longer and offers protection against a wider range of hazardous substances.
- Approved for food processing according to EU 1935/2004
- Food grade through RAL approval
- Silicone free glove surface
- Meets the requirements of EN 455
- Medical device according to directive 93/42/EWG
- Free from natural latex
- Resistant against a multitude of cytostatic drugs virus-resistant according to ASTM 1671:2007
- High tactility
- Excellent wearing comfort
- Increased tear resistance
- Improved grip
- Good chemical resistance
- Snug fit
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Certifications/Compliance | ASTM 1671:2007 |
Certifications/Compliance (Europe) | Approved for food processing according to EU 1935/2004. EC Category PSA 3 |
Color | Blue |
Latex Free | Yes |
Material | Nitrile |
Powdered | Powder Free |
Size | 6 |
Thickness (Metric) | 0.2 mm |
Quantity | 50/Pk |
Resistant To | Variety of cytotoxic agents, virus-resistant to ASTM F1671: 2007 |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'