missing translation for 'onlineSavingsMsg'
Learn More

Honeywell KCL™ Dermatril™ P 743 Gloves

Product Code. 15320392
Click to view available options
Size:
6
7
8
9
10
11
Unit Size:
Pack of 50

Product Code. 15320392

Brand: Honeywell KCL™ 074308082C

Only null left
Add to basket
Add to basket

Honeywell KCL™ Dermatril™ P 743 Gloves feature chemical and mechanical resistance and are approved for food processing.

  • Disposable protective glove according to EN 374
  • The chemical as well as the mechanical resistance are increased compared to the Dermatril™ 740
  • Can be used longer and offers protection against a wider range of hazardous substances.
  • Approved for food processing according to EU 1935/2004
  • Food grade through RAL approval
  • Silicone free glove surface
  • Meets the requirements of EN 455
  • Medical device according to directive 93/42/EWG
  • Free from natural latex
  • Resistant against a multitude of cytostatic drugs virus-resistant according to ASTM 1671:2007
  • High tactility
  • Excellent wearing comfort
  • Increased tear resistance
  • Improved grip
  • Good chemical resistance
  • Snug fit
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance ASTM 1671:2007
Certifications/Compliance (Europe) Approved for food processing according to EU 1935/2004. EC Category PSA 3
Color Blue
Latex Free Yes
Material Nitrile
Powdered Powder Free
Size 8
Thickness (Metric) 0.2 mm
Quantity 50/Pk
Resistant To Variety of cytotoxic agents, virus-resistant to ASTM F1671: 2007
Length (Metric) 270 to 290 mm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.