missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Desiccator with Glass Knob
Made from Simax 3.3 borosilicate glass. Fisherbrand Desiccator with Glass Knob is designed for variety of vacuum work.
Brand: Fisherbrand™ 632415280253
Description
- Made from SIMAX 3.3 borosilicate glass
- Designed for variety of vacuum work
- Used for desiccation of samples, storage of materials sensitive to humidity
Specifications
ISO 13 130, DIN 12 491 | |
Desiccator | |
363 mm | |
320 mm |
Simax Borosilicate Glass 3.3 | |
Glass knob | |
239 mm |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction