missing translation for 'onlineSavingsMsg'
Learn More

Desmin Antibody (rDES/1711), DyLight 550, Novus Biologicals™

Mouse Monoclonal Antibody

Brand:  Bio-Techne NBP3-14038R

Product Code. 18317442

  • £474.00 / 0.10mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Desmin Monoclonal antibody specifically detects Desmin in Human, Mouse, Rat, Chicken, Hamster samples. It is validated for Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications

Specifications

Desmin
Monoclonal
DyLight 550
CMD1IFLJ41013, CSM1, CSM2, desmin, FLJ12025, FLJ39719, FLJ41793, intermediate filament protein
Recombinant full-length human desmin protein (Uniprot: P17661)
0.1 mL
Cell Biology, Cellular Markers
1674
Store at 4°C in the dark.
IgG1 κ
Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
rDES/1711
50mM Sodium Borate
Mouse
Protein A or G purified
RUO
Primary
Human, Mouse, Rat, Chicken, Hamster
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.