missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Lab Vision™ Desmin (Muscle Cell Marker) Ab-1, Mouse Monoclonal Antibody

Specifically detect desmin in human, baboon, monkey, cow, cat, dog, hamster, rat and chicken samples.

Brand:  Epredia™ MS-376-S

344.08 GBP valid until 2024-12-31
Use promo code "21628" to get your promotional price.


 View more versions of this product

Product Code. 12641227

  • £405.00 / 1mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Desmin is an intermediate filament protein of both smooth and striated muscles. Antibody to desmin reacts with striated (skeletal and cardiac) as well as smooth muscle cells. In skeletal and cardiac muscles, the staining is confined to the Z-bands giving a characteristic striated appearance. Anti-desmin antibody is useful in identification of tumors of myogenic origin. It reacts with leiomyosarcomas (smooth muscle) as well as rhabdomyosarcomas (striated muscle).

Host Species: Mouse

Clone: D33

Isotype: IgG1/κ

Species Reactivity: Human, Baboon, Monkey, Cow, Cat, Dog, Hamster, Rat and Chicken. Others not known.

Epitope: Not determined

Immunogen: Desmin from human muscle

Molecular Weight: 53 kDa

Positive Control: Muscle or Sarcoma

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Immunofluorescence
Specifications

Specifications

Desmin (Muscle Cell Marker) Ab-1
Monoclonal
Unconjugated
Desmin from human muscle
IVD
Primary
Concentrated
Immunofluorescence, Immunohistochemistry (Paraffin)
D33
Mouse
1 mL
Muscle Biology
Bovine, Canine, Chicken, Feline, Hamster, Human, Non-human Primate, Rat
IgG2a κ
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.