Learn More
Dess-Martin periodinane, 15 wt.% solution in dichloromethane, Thermo Scientific Chemicals
Dess-Martin periodinane, 10 to 15%, C13H13IO8, CAS Number-87413-09-0, 75-09-2, dess martin, dess-martin, dess-martinperiodinane, dess-martin periodinane, 1,1,1-triacetoxy-1,1-dihydro-1,2-benziodoxol-3 1h-one, dess martin periodinane, dess-martin periodane, dess-martin reagent, triacetoxyperiodinane
£48.04 - £528.63
Specifications
Chemical Name or Material | Dess-Martin periodinane |
---|---|
CAS | 87413-09-0, 75-09-2 |
Molecular Formula | C13H13IO8 |
MDL Number | MFCD00130127 |
Density | 1.3620g/mL |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10562861
|
Thermo Scientific Acros
333110500 |
50 mL |
£207.03
50mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15284704
|
Thermo Scientific Acros
333112500 |
250 mL |
£528.63
250mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15394978
|
Thermo Scientific Acros
333110100 |
10 mL |
£48.04
10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
oxidation reagentSpecifications
Dess-Martin periodinane | |
C13H13IO8 | |
1.3620g/mL | |
NKLCNNUWBJBICK-UHFFFAOYSA-N | |
424.14 | |
424.15 |
87413-09-0, 75-09-2 | |
MFCD00130127 | |
dess-martin periodinane, triacetoxyperiodinane, 1,1,1-triacetoxy-1,1-dihydro-1,2-benziodoxol-3 1h-one, dess-martin reagent, dess-martinperiodinane, dess-martin, dess martin periodinane, dess martin, dess-martin periodane | |
CC(=O)O[I]1(OC(C)=O)(OC(C)=O)OC(=O)C2=CC=CC=C12 | |
159087 | |
1.362 |
Safety and Handling
GHS H Statement:
Suspected of causing cancer.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
May intensify fire; oxidizer.
GHS P Statement:
Use personal protective equipment as required.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Danger
Recommended Storage : Product may develop some turbidity or precipitate
RUO – Research Use Only