Learn More
Cytiva DeStreak Rehydration Solution
Highly suitable for both analytical and preparative gels. DeStreak Rehydration Solution improves reproducibility and quality of 2-D gels by preventing streaking.
Brand: Cytiva 17-6003-18
61.52 GBP valid until 2024-12-31
Use promo code "21631" to get your promotional price.
View more versions of this product
Description
The appearance of streaks that distort 2-D electrophoresis maps is a common problem, occurring most frequently when running gels that contain regions greater than pH 7.0. Increased sample load, increased length of the IPG strip, or using a narrower pH gradient worsen the problem. Extra spots on 2-D gels, caused by nonspecific oxidation of proteins, is another difficulty encountered when running gels containing basic regions. Both streaking and nonspecific oxidation result in poorly resolved protein patterns and reduced reproducibility between electrophoresis runs. DeStreak Rehydration Solution effectively eliminates nonspecific oxidation by maintaining protein thiol groups in a single oxidation state, irrespective of sample load, pH range, or run length. Protein patterns are stabilized in IPG strips of any length and pH gradient ensuring the same stable and reproducible pattern in every analysis. The effectiveness of DeStreak Rehydration Solution is maintained at the high sample loads and long run times required for preparative gel electrophoresis.
- Eliminates extra spots caused by nonspecific oxidation of proteins
- Stabilizes proteins throughout entire 2-D runs, ensuring simplified, well-resolved protein patterns
- Suitable for both analytical and preparative gels
- Contains optimized concentrations of urea, thiourea, CHAPS, and DeStreak Reagent
Order Info
The solution is ready for use after addition of the appropriate IPG Buffer. DeStreak Reagent is also available separately in a 1mL pack size.
Specifications
Improves reproducibility and quality of 2D Gels by preventing streaking | |
Destreak Reagent | |
>7 |
1 mL | |
Optimized concentrations of urea, thiourea, CHAPS and Destreak reagent | |
2°C to 8°C |
Safety and Handling
- Danger
Hazard Category
- Acute toxicity Category 3
Hazard Statement
- H301-Toxic if swallowed.
Precautionary Statement
- P270-Do not eat, drink or smoke when using this product.
- P264-Wash thoroughly after handling.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P330-Rinse mouth.
- P405-Store locked up.
- P501-Dispose of contents/container to .
Your input is important to us. Please complete this form to provide feedback related to the content on this product.