Learn More
Cole-Parmer™ Detachable Electrode Cable
Features BNC connection for compatibility with most pH meters and controllers
Brand: Cole-Parmer™ 05662-56
Code : NEW
Additional Details : Weight : 0.06000kg
Description
- Electrode cable, detachable, 10-ft, BNC
- Compatible with most pH meters and controllers
Specifications
Cole-Parmer Electrode Cable, Detachable, 10-ft, BNC | |
BNC Connection | |
10 ft. | |
3 m |