Learn More
Detritylation Reagent, Honeywell Burdick & Jackson™
BioSyn™, suitable for use with DNA/RNA synthesis instruments, 3%Trichloroacetic Acid in Dichloromethane (withv)
Brand: Honeywell Fluka BR605-4x2.5L
Additional Details : Weight : 1.32990kg
Description
Burdick & Jackson™ BioSyn™ reagents & solvents for DNA/RNA synthesis
- Honeywell Burdick & Jackson™ BioSyn™ Solvents and Reagents are specifically processed and purified to exacting specifications, ensuring outstanding synthesis efficiency
- ideal for oligonucleotide synthesis regardless of the application
- All solvents and reagents are filtered (0.2-micron for oxidation reagents and 0.1-micron for all other reagents) to minimize particulate contamination
Specifications
75-09-2,76-03-9 | |
40°C | |
Glass Bottle | |
Deblocking reagents |
3.00% | |
4 x 2.5 L | |
Liquid, clear |