Learn More
Fisherbrand™ Deuterium Lamps
Designed for compatibility with a variety of instruments. Fisherbrand™ Deuterium Lamps are individually tested, certified, and guaranteed to the same specifications as OEM.
Brand:  Fisherbrand™ 11958-19
610.85 GBP valid until 2025-04-30
Use promo code "24163" to get your promotional price.
 View more versions of this product
Product Code. 16250942
  • £643.00 / Each
Explore more special offers
Description
Description
  • Produced in state of the art manufacturing facilities approved by lamp manufacturers
  • Full traceability of manufactured products
  • Lamps available for all major instruments
Specifications
Specifications
UV
1 mm
1 Ea.
400/185 nm
PE Lambda 2 to 45 800 900 Bio, 55X Series LC480
Deuterium Lamp
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Fisherbrand™ Deuterium Lamps > PE Lambda 2 to 45 800 900 Bio, 55X Series LC480 Detectors