Learn More
Fisherbrand™ Deuterium Lamps
Designed for compatibility with a variety of instruments. Fisherbrand™ Deuterium Lamps are individually tested, certified, and guaranteed to the same specifications as OEM.
Brand:  Fisherbrand™ 11957-73
 View more versions of this product
Product Code. 16260942
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • Produced in state of the art manufacturing facilities approved by lamp manufacturers
  • Full traceability of manufactured products
  • Lamps available for all major instruments
Specifications
Specifications
Deuterium Lamp
400/185 nm
Shimadzu UV series Spectrophotometer & BioSpec-mini
UV
1 mm
1 Ea.