Learn More
Fisherbrand™ Deuterium Lamps
Designed for compatibility with a variety of instruments. Fisherbrand™ Deuterium Lamps are individually tested, certified, and guaranteed to the same specifications as OEM.
Brand:  Fisherbrand™ 11958-21
649.75 GBP valid until 2024-12-31
Use promo code "21658" to get your promotional price.
 View more versions of this product
Product Code. 16270942
  • £738.00 / Each
Explore more special offers
Description
Description
  • Produced in state of the art manufacturing facilities approved by lamp manufacturers
  • Full traceability of manufactured products
  • Lamps available for all major instruments
Specifications
Specifications
Deuterium Lamp
400/160 nm
Varian Prostar 330 DAD
UV
1 mm
1 Ea.