Learn More
Fisherbrand™ Deuterium Lamps
Designed for compatibility with a variety of instruments. Fisherbrand™ Deuterium Lamps are individually tested, certified, and guaranteed to the same specifications as OEM.
Brand:  Fisherbrand™ 11958-23
694.45 GBP valid until 2025-04-30
Use promo code "24163" to get your promotional price.
 View more versions of this product
Product Code. 16290942
  • £731.00 / Each
Explore more special offers
Description
Description
  • Produced in state of the art manufacturing facilities approved by lamp manufacturers
  • Full traceability of manufactured products
  • Lamps available for all major instruments
Specifications
Specifications
UV
0.5 mm
1 Ea.
400/185 nm
Waters Acquity 2489/2998
Deuterium Lamp
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Fisherbrand™ Deuterium Lamps > Waters Acquity 2489/2998 Detectors