missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Deuterium oxide, for NMR, 99.8 atom % D, Thermo Scientific Chemicals
£21.10 - £888.00
Chemical Identifiers
CAS | 7789-20-0 |
---|---|
Molecular Formula | H2O |
Molecular Weight (g/mol) | 20.03 |
MDL Number | MFCD00044636 |
InChI Key | XLYOFNOQVPJJNP-ZSJDYOACSA-N |
Synonym | deuterium oxide, water-d2, heavy water, deuterated water, dideuterium oxide, heavy water-d2, heavy water d2o, water sup 2-h2, deuterium oxide usan, water, heavy d2-o |
PubChem CID | 24602 |
ChEBI | CHEBI:41981 |
IUPAC Name | [(²H)oxy](²H) |
SMILES | [2H]O[2H] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10584321
|
Thermo Scientific Acros
166300100 |
10 mL |
£21.10
10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16120891
|
Thermo Scientific Acros
166300250 |
25 mL |
£39.60
25mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10255880
|
Thermo Scientific Acros
166301000 |
100 mL |
£140.00
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16130891
|
Thermo Scientific Acros
166305000 |
500 mL |
£550.00
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10220953
|
Thermo Scientific Acros
166300010 |
1 L |
£888.00
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For NMRChemical Identifiers
7789-20-0 | |
20.03 | |
XLYOFNOQVPJJNP-ZSJDYOACSA-N | |
24602 | |
[(²H)oxy](²H) |
H2O | |
MFCD00044636 | |
deuterium oxide, water-d2, heavy water, deuterated water, dideuterium oxide, heavy water-d2, heavy water d2o, water sup 2-h2, deuterium oxide usan, water, heavy d2-o | |
CHEBI:41981 | |
[2H]O[2H] |
Specifications
Liquid | |
1.1050g/mL | |
Glass Bottle | |
1.105 | |
7789-20-0 | |
101.4°C | |
D2O | |
15, 2942 | |
Solubility in water: miscible, exothermic reaction. | |
[2H]O[2H] | |
20.03 | |
CHEBI:41981 | |
NMR |
Colorless | |
99.75% min. | |
1.328 | |
1.25 mPa.s (20°C) | |
3.8°C | |
H2O | |
MFCD00044636 | |
deuterium oxide, water-d2, heavy water, deuterated water, dideuterium oxide, heavy water-d2, heavy water d2o, water sup 2-h2, deuterium oxide usan, water, heavy d2-o | |
XLYOFNOQVPJJNP-ZSJDYOACSA-N | |
[(²H)oxy](²H) | |
24602 | |
20.02 | |
Deuterium oxide, 99.8 atom % D |
Safety and Handling
HYGROSCOPIC
missing translation for 'einecsNumber' : 232-148-9
missing translation for 'rtecsNumber' : ZC0230000
missing translation for 'tsca' : TSCA
RUO – Research Use Only