missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Deuterosulfuric acid, 96% min in D{2}O, 99.5% (Isotopic), Thermo Scientific Chemicals
Click to view available options
Quantity:
50 g
Description
Deuterated sulfuric acid when treated with benzene, the deuterium is used in order to observe the reaction course by NMR spectroscopy, deuterated sulfuric acid results in an electrophilic aromatic substitution of hydrogen by deuterium.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Physical Form | Liquid |
Density | 1.86 |
Refractive Index | 1.4175 |
Sensitivity | Moisture |
Quantity | 50 g |
CAS | 13813-19-9 |
Boiling Point | 290°C |
Assay Percent Range | <96% in D2O, 99.5% (Isotopic) |
Molecular Formula | H2O4S |
MDL Number | MFCD00003542 |
Show More |
Safety and Handling
Product Identifier
- Deuterosulfuric acid, 96-98% in D2O
- Danger
- Skin corrosion/irritation Category 1 A
- H314-Causes severe skin burns and eye damage.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- MIXTURE LIST-Contains : Sulfuric acid-d2
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'