missing translation for 'onlineSavingsMsg'
Learn More

Dexamethasone, Tocris Bioscience™

Product Code. 30001202
Click to view available options
Quantity:
100 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Anti-inflammatory glucocorticoid

Glucocorticoid; anti-inflammatory. Reduces levels of activated NF-κB in immature dendritic cells (DCs) and inhibits differentiation into mature DCs. Induces differentiation of human mesenchymal stem cells (MSCs). Also used in the generation of liver organoids. Induces autophagy in acute lymphoblastic leukemia (ALL) cell lines.

Tocris products are for biomedical research use only. They are not intended for human or veterinary use.

Specifications

CAS 50-02-2
Molecular Formula C22H29FO5
MDL Number MFCD00064136
Quantity 100 mg
Synonym dexamethasone, decadron, dexamethazone, maxidex, decaspray, desametasone, hexadecadrol, dexacort, dexasone, hexadrol
InChI Key UREBDLICKHMUKA-CXSFZGCWSA-N
SMILES C[C@@H]1C[C@H]2[C@@H]3CCC4=CC(=O)C=C[C@]4(C)[C@@]3(F)[C@@H](O)C[C@]2(C)[C@@]1(O)C(=O)CO
IUPAC Name (1R,2R,3aS,3bS,9aS,9bR,10S,11aS)-9b-fluoro-1,10-dihydroxy-1-(2-hydroxyacetyl)-2,9a,11a-trimethyl-1H,2H,3H,3aH,3bH,4H,5H,7H,9aH,9bH,10H,11H,11aH-cyclopenta[a]phenanthren-7-one
Molecular Weight (g/mol) 392.47
PubChem CID 5743
ChEBI CHEBI:41879
Formula Weight 392.47
Percent Purity >98%
Chemical Name or Material Dexamethasone
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'