missing translation for 'onlineSavingsMsg'
Learn More

Honeywell Safety Products™ Dexgrip Carded Gloves

Product Code. 12655193
Click to view available options
Size:
7
8
9
This item is not returnable. View return policy
This item is not returnable. View return policy

Comfortable, hand conforming and snug, this knit glove provides both breathability and grip for handling of damp or non-greasy parts. Honeywell Safety Products™ Dexgrip Carded Gloves come in effective, single-pair packaging designed for a self-service environment.

  • Knitted cotton/polyamide grey glove.
  • Palm and fingertips coated in blue, crinkled latex for abrasion and perforation resistance as well as excellent grip.
  • Conforms snugly and provides good dexterity.
  • Cotton thread absorbs perspiration.
  • Ventilated back provides breathability.
  • Product emphasized in effective, single-pair packaging designed for a self-service environment.
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance CE CATEGORY II Intermediary risks, EN 420-2003 Basic standard, EN 388-2003 Mechanical risks, EN 407-2004 Thermal risks, EC Category PPE 2, Quality Assurance ISO 9001 / 2000, EC Certificate Number 0075/797/162/12/08/0519, EC Attestation Number 0075/797/162/12/08/0519
Classification Dexterity: 5
Coated Area Palm, Fingertips
Description Gauge 10
Coating Material Crinkled Latex
Color Gray, Blue
Cut Resistance Rating 1
Material Polyamide, Latex, Cotton
Product Type Gloves
Size 7
Thickness (Metric) 1.5 to 1.8 mm
For Use With (Application) Agriculture, Building, Construction, Glass, Green Spaces, Logistics, Transportation and Wood industries
Packaging Type Single pair
Quantity 10Pair/PK
Tear Rating 4
Length (Metric) 26 cm
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'