Learn More
Honeywell™ DEXGRIP LIGHT CARDED Gloves
Click to view available options
Size: 8
Description
- Knitted glove cotton/polyamide blue
- Palm and fingertips coated in blue color crinkled latex
- Knitted wrist
- The knitting technique helps to obtain a glove that conforms snugly to the hand and good dexterity
- The glove offers maximum comfort and added sturdiness
- The tight knit (gauge 13) gives the glove perfect support, limits the penetration of dirt and ensures a good touch
- The cotton thread absorbs perspiration and thus provides greater comfort
- Its ultra fine crinkled latex coating offers maximum dexterity while providing mechanical resistance
- The grip offers high performance even in a damp environment
- Product delivered in a clear and effective packaging designed for a self-service environment, which provides the needed information to easily make the right protection choice
- It includes pictograms to easily recognize the protection range as well as hazards identification and product advantages
- Customers can easily recognize which personal protective equipment to choose, depending on their work environment and the risks they face
- As a result, they become autonomous and can be sure they are making the right protection choice
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Certifications/Compliance | CE CATEGORY : II intermediary risks In accordance with standards EN 420-2003 Basic standard. EN 388-2003 Mechanical risks |
Description | 13 gauge |
Color | Blue |
Material | Cotton, Polyamide |
Size | 8 |
Length (Metric) | 26 cm |