missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DEXI Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
£135.00 - £349.00
Specifications
Antigen | DEXI |
---|---|
Concentration | 0.29 mg/mL |
Applications | Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16863644
|
Proteintech
14811-1-AP-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16853644
|
Proteintech
14811-1-AP-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
DEXI (dexamethasone-induced protein), also known as MYLE, is a 95 amino acid protein belonging to the DEXI family.Specifications
DEXI | |
Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
O95424, Q9WUQ7 | |
28955, 497857, 58239 | |
DEXI Fusion Protein Ag6535 | |
Primary | |
-20°C | |
DEXI |
0.29 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Dexamethasone induced protein, DEXI, MYLE, Protein MYLE | |
DEXI | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |