missing translation for 'onlineSavingsMsg'
Learn More

Dexmedetomidine hydrochloride, Tocris Bioscience™

Product Code. 30001441
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Potent, highly selective α2 agonist. Active isomer of medetomidine (Cat. No. 5160)

Dexmedetomidine hydrochloride is an active isomer of medetomidine, a potent, highly selective α2-adrenoceptor agonist (Ki values are 1.08 and 1750 nM for α2- and α1-adrenoceptors respectively). Displays greater selectivity over α1-adrenoceptors than Clonidine (Cat. No. 0690) and UK 14,304 (1620-, 220- and 300-fold respectively). Active in vivo; displays hypotensive, bradycardic, sedative, anxiolytic, hypothermic and analgesic effects.

Racemate also available.

Chemical Identifiers

CAS 145108-58-3
Molecular Formula C13H17ClN2
Molecular Weight (g/mol) 236.743
InChI Key VPNGEIHDPSLNMU-MERQFXBCSA-N
Synonym dexmedetomidine hydrochloride, dexmedetomidine hcl, precedex, 4-s-alpha,2,3-trimethylbenzyl imidazole monohydrochloride, unii-1018wh7f9i, dexmedetomidine hcl precedex, s-5-1-2,3-dimethylphenyl ethyl-1h-imidazole hydrochloride, dexdor, 4-1s-1-2,3-dimethylphenyl ethyl-1h-imidazole hydrochloride
PubChem CID 6918081
ChEBI CHEBI:31472
IUPAC Name 5-[(1S)-1-(2,3-dimethylphenyl)ethyl]-1H-imidazole;hydrochloride
SMILES CC1=C(C(=CC=C1)C(C)C2=CN=CN2)C.Cl

Specifications

Quantity 10 mg
Formula Weight 236.74
Percent Purity 98%
Chemical Name or Material Dexmedetomidine hydrochloride
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'