missing translation for 'onlineSavingsMsg'
Learn More
Dextran Sulfate Sodium Salt, Thermo Scientific Chemicals Product Code.: 15885118

Dextran Sulfate Sodium Salt, Thermo Scientific Chemicals

Product Code. 15885118 Shop All Thermo Scientific Products
50 g, Plastic bottle
Click to view available options
Quantity:
10 g
50 g
100 g
Packaging:
Plastic bottle
This item is not returnable. View return policy

Product Code. 15885118

Brand: Thermo Scientific J14489.18

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 9011-18-1
Molecular Formula (C6H7O5)m(C6H7O5)n
Molecular Weight (g/mol) 40,000
MDL Number MFCD00081551
InChI Key BLFLLBZGZJTVJG-UHFFFAOYSA-N
Synonym benzocaine, ethyl aminobenzoate, ethyl p-aminobenzoate, americaine, anesthesin, anaesthesin, ethoform, norcaine, orthesin, parathesinShow MoreShow Less
PubChem CID 2337
ChEBI CHEBI:116735
IUPAC Name ethyl 4-aminobenzoate
SMILES *-OC[C@H]1O[C@H](OC[C@H]2O[C@H](-*)[C@H](O[*])[C@@H](O[*])[C@@H]2O[*])[C@H](O[*])[C@@H](O-*)[C@@H]1O[*]

Specifications

Physical Form Powder
Quantity 50 g
Formula Weight 40,000- 50,000
Packaging Plastic bottle
Chemical Name or Material Dextran Sulfate Sodium Salt

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number230228A Certificate TypeCertificate of Analysis Date11/06/2024 Catalog NumberJ14489.22,J14489.18,J14489.09
Lot Number230228 Certificate TypeCertificate of Analysis Date09/05/2023 Catalog NumberJ14489.22,J14489.18,J14489.09
Lot Number187742 Certificate TypeCertificate of Analysis Date27/11/2022 Catalog NumberJ14489.22,J14489.18,J14489.09
Lot Number178017 Certificate TypeCertificate of Analysis Date27/11/2022 Catalog NumberJ14489.22,J14489.18,J14489.09
Lot Number207446 Certificate TypeCertificate of Analysis Date24/06/2022 Catalog NumberJ14489.22,J14489.18,J14489.09

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Dextran Sulfate Sodium Salt, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.