missing translation for 'onlineSavingsMsg'
Learn More

Dextrin, Thermo Scientific Chemicals

Product Code. 10351441
Click to view available options
Quantity:
100 g
500 g
Packaging:
Plastic bottle

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9004-53-9
Molecular Formula C18H32O16
Molecular Weight (g/mol) 504.438
MDL Number MFCD00081554
InChI Key FYGDTMLNYKFZSV-MRCIVHHJSA-N
Synonym dextrin, dextrins, fortodex, dextrid, corn dextrin, dextrina bianca, british gum, crystal gum, dextrin 3, caloreen
PubChem CID 62698
IUPAC Name (3R,4S,5S,6R)-2-[(2R,3S,4R,5R)-4,5-dihydroxy-2-(hydroxymethyl)-6-[(2R,3S,4R,5R,6S)-4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxyoxan-3-yl]oxy-6-(hydroxymethyl)oxane-3,4,5-triol
SMILES C(C1C(C(C(C(O1)OC2C(OC(C(C2O)O)OC3C(OC(C(C3O)O)O)CO)CO)O)O)O)O

Specifications

Color White to Yellow
Physical Form Crystalline Powder
Linear Formula (C6H10O5)x
Merck Index 15, 2955
Solubility Information Solubility in water: soluble in hot water. Other solubilities: insoluble in ether and alcohols
Quantity 500 g
Formula Weight 162.07
Infrared Spectrum Authentic
Packaging Plastic bottle
Chemical Name or Material Dextrin

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'