Learn More
Dextrose Anhydrous (Crystalline Granules/Molecular Biology), Fisher BioReagents
£33.05 - £50.55
Chemical Identifiers
CAS | 50-99-7 |
---|---|
Molecular Formula | C6H12O6 |
Molecular Weight (g/mol) | 180.156 |
InChI Key | GZCGUPFRVQAUEE-SLPGGIOYSA-N |
Synonym | 2r,3s,4r,5r-2,3,4,5,6-pentahydroxyhexanal, d +-glucose, aldehydo-d-glucose, glucose, anhydrous, dextrose, anhydrous, d-glucose, anhydrous, glucosteril, anhydrous dextrose, dextrose solution, d-glucose in linear form |
PubChem CID | 107526 |
ChEBI | CHEBI:42758 |
IUPAC Name | (2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanal |
SMILES | C(C(C(C(C(C=O)O)O)O)O)O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10051793
|
Fisher BioReagents
BP350-500 |
500 g | Glass Bottle |
£33.05
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10764534
|
Fisher BioReagents
BP350-1 |
1 kg | Poly Bottle |
£50.55
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Dextrose is often used as a cell culture additive.Chemical Identifiers
50-99-7 | |
180.156 | |
2r,3s,4r,5r-2,3,4,5,6-pentahydroxyhexanal, d +-glucose, aldehydo-d-glucose, glucose, anhydrous, dextrose, anhydrous, d-glucose, anhydrous, glucosteril, anhydrous dextrose, dextrose solution, d-glucose in linear form | |
CHEBI:42758 | |
C(C(C(C(C(C=O)O)O)O)O)O |
C6H12O6 | |
GZCGUPFRVQAUEE-SLPGGIOYSA-N | |
107526 | |
(2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanal |
Specifications
50-99-7 | |
White | |
≥99.8 % | |
HOCH2(CHOH)4CHO | |
52.5 to 53.0° (+ or -) | |
0.005% max. Insoluble Matter | |
C(C(C(C(C(C=O)O)O)O)O)O | |
180.156 | |
CHEBI:42758 | |
Negligible | |
DNase-, RNase- and Protease-Free | |
Dextrose Anhydrous |
146.1°C | |
5.9 | |
C6H12O6 | |
15, 4495 | |
2r,3s,4r,5r-2,3,4,5,6-pentahydroxyhexanal, d +-glucose, aldehydo-d-glucose, glucose, anhydrous, dextrose, anhydrous, d-glucose, anhydrous, glucosteril, anhydrous dextrose, dextrose solution, d-glucose in linear form | |
GZCGUPFRVQAUEE-SLPGGIOYSA-N | |
(2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanal | |
107526 | |
180.08 | |
≥99.8% | |
Solid |
Safety and Handling
CAUTION!
Emergency Overview
May cause skin, eye, and respiratory tract irritation. Use personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur. Do not induce vomiting. Obtain medical attention. Obtain medical attention. .
NFPA
Health:1
Flammability:0
Instability:0
Recommended Storage : RT
Your input is important to us. Please complete this form to provide feedback related to the content on this product.