missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Applied Biosystems™ dGTP BigDye™ Terminator v1.0 & v3.0 5X Sequencing Buffer
This sequencing buffer is optimized for use with BigDye™ Terminator v1.0 or v3.0 cycle sequencing kits.
Brand: Applied Biosystems™ 4305605
Additional Details : Weight : 0.01000kg
Description
This sequencing buffer is optimized for use with BigDye™ Terminator v1.0 or v3.0 cycle sequencing kits.Order Info
1.2mL
Specifications
Sequencing Buffer | |
Store in freezer (-5 to -30°C). | |
GT-Rich or Difficult Template Sequencing | |
Sequencing Buffer | |
Dry Ice |
5 X | |
GeneAmp 9700, Veriti Thermal Cycler, 3500 Genetic Analyzer, 3730xl DNA Analyzer, 3130xl Genetic Analyzer, 3500xL Genetic Analyzer, 3730 DNA Analyzer | |
BigDye™ Terminator, BigDye™ | |
1.2 mL | |
Liquid |
For Research Use Only. Not for use in diagnostic procedures.