Learn More
Thermo Scientific™ DH5α Competent Cells; High Efficiency
High efficiency, chemically competent E. coli cells ideal for construction of gene banks or generation of cDNA libraries using plasmid-derived vectors
Brand: Thermo Scientific™ EC0112
81.33 GBP valid until 2024-06-29
Use promo code "21620" to get your promotional price.
Additional Details : Weight : 0.04500kg
Description
- The φ80dlacZΔM15 marker provides α-complementation of the β-galactosidase gene from pUC or similar vectors to allow blue/white colony screening on bacterial agar plates containing Bluo-Gal or X-Gal
- RecA1 and endA1 mutations in DH5α cells improve insert stability and quality of the extracted plasmid DNA
- High transformation efficiency: >1x109 cfu/μg pUC19 DNA
- Suitable for routine and high-throughput cloning applications
- Genetic markers that allow for blue/white colony screening
- Convenient two reactions per vial packaging
- S.O.C. Outgrowth medium included
- DH5α Competent Cells are suitable for variety of applications requiring high transformation efficiency:
- Efficient DNA cloning derived from PCR, cDNA-generation reactions
- recA1 marker facilitates working with difficult to transform DNA
- Ideal for generation of cDNA libraries using plasmid-derived vectors
- Site-directed mutagenesis
- Genotype: F– φ80lacZΔ M15 Δ (lacZYA-argF) U169 recA1 endA1 hsdR17 (rK– mK+) phoA supE44 λ- thi–1 gyrA96 relA1
Specifications
Chemically Competent | |
Dry Ice (-80°C) | |
E. coli | |
Cloning |
Lacks F' Episome | |
DH5α | |
Tube | |
20 Reactions |