Learn More
DHX16 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 11021-1-AP-20UL
 View more versions of this product
Product Code. 16887803
  • £135.00 / 20µL
Explore more special offers
Description
Description
DEAD box proteins, characterized by the conserved motif Asp-Glu-Ala-Asp (DEAD), are putative RNA helicases. They are implicated in a number of cellular processes involving alteration of RNA secondary structure such as translation initiation, nuclear and mitochondrial splicing, and ribosome and spliceosome assembly. Based on their distribution patterns, some members of this family are believed to be involved in embryogenesis, spermatogenesis, and cellular growth and division. This gene encodes a DEAD box protein, which is a functional homolog of fission yeast Prp8 protein involved in cell cycle progression. This gene is mapped to the MHC region on chromosome 6p21.3, a region where many maligt, genetic and autoimmune disease genes are linked. Three transcript variants encoding different isoforms have been found for this gene.
Specifications
Specifications
DHX16
Polyclonal
Unconjugated
DHX16
ATP dependent RNA helicase 3, DBP2, DDX16, DEAH box protein 16, DHX16, KIAA0577, PRO2014, PRP8
Rabbit
Antigen Affinity Chromatography
RUO
69192, 8449
-20°C
Liquid
Western Blot, Immunofluorescence, Immunohistochemistry (Paraffin), Immunocytochemistry
0.35 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
O60231
DHX16
DHX16 Fusion Protein Ag1482
20 μL
Primary
Mouse, Human