missing translation for 'onlineSavingsMsg'
Learn More

Di-Iso-Propyl Ether, Certified AR for Analysis, Stabilised with BHT, Fisher Chemical™

Product Code. 10082923
Click to view available options
Quantity:
1 L
2.5 L
5 L
25 L
Packaging:
Amber glass bottle
Metal can
This item is not returnable. View return policy
This item is not returnable. View return policy

C6H14O, CAS Number-108-20-3, 2,2'-oxydipropane, 2-isopropoxypropane, bis isopropyl ether, diisopropylether, diisopropyl ether, diisopropyl oxide, ether, isopropyl, isopropylether, isopropyl ether, propane, 2,2'-oxybis, 2.5L, 68 deg.C, Colorless, 102.18g/mol, ZAFNJMIOTHYJRJ-UHFFFAOYSA-N

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108-20-3
Molecular Formula C6H14O
Molecular Weight (g/mol) 102.177
MDL Number 8880
InChI Key ZAFNJMIOTHYJRJ-UHFFFAOYSA-N
Synonym diisopropyl ether, isopropyl ether, propane, 2,2'-oxybis, 2-isopropoxypropane, diisopropyl oxide, diisopropylether, ether, isopropyl, 2,2'-oxydipropane, isopropylether, bis isopropyl ether
PubChem CID 7914
IUPAC Name 2-propan-2-yloxypropane
SMILES CC(C)OC(C)C

Specifications

Melting Point -85.5°C
Color Colorless
Boiling Point 68°C
Packaging Metal can
Quantity 25 L
Vapor Density 1.42
Formula Weight 102.18g/mol
Physical Form Liquid
compliance-icons compliance-icons
Product Identifier
  • Isopropyl ether
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H336-May cause drowsiness or dizziness.
  • EUH019-May form explosive peroxides.
  • EUH066-Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P233-Keep container tightly closed.
  • P240-Ground/bond container and receiving equipment.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'