missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Di-Iso-Propyl Ether, Certified AR for Analysis, Stabilised with BHT, Fisher Chemical™
Click to view available options
Quantity:
1 L
2.5 L
5 L
25 L
Packaging:
Amber glass bottle
Metal can
Description
C6H14O, CAS Number-108-20-3, 2,2'-oxydipropane, 2-isopropoxypropane, bis isopropyl ether, diisopropylether, diisopropyl ether, diisopropyl oxide, ether, isopropyl, isopropylether, isopropyl ether, propane, 2,2'-oxybis, 2.5L, 68 deg.C, Colorless, 102.18g/mol, ZAFNJMIOTHYJRJ-UHFFFAOYSA-N
Specifications
Specifications
Melting Point | -85.5°C |
Color | Colorless |
Boiling Point | 68°C |
Packaging | Metal can |
Quantity | 5 L |
Vapor Density | 1.42 |
Formula Weight | 102.18g/mol |
Physical Form | Liquid |
Safety and Handling
Product Identifier
- Isopropyl ether
- Danger
- Flammable liquid Category 2
- Specific target organ toxicity Category 3
- H225-Highly flammable liquid and vapour.
- H336-May cause drowsiness or dizziness.
- EUH019-May form explosive peroxides.
- EUH066-Repeated exposure may cause skin dryness or cracking.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P233-Keep container tightly closed.
- P240-Ground/bond container and receiving equipment.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'