missing translation for 'onlineSavingsMsg'
Learn More

Di-Lauroyl Peroxide, Pure, Fisher Chemical™

Product Code. 10548620 Shop All Fisher Chemical Products
Click to view available options
Quantity:
5 x 1 g
100 g
Packaging:
Plastic powder jar
Vial
Unit Size:
1 pack
100g
This item is not returnable. View return policy

Product Code. 10548620

missing translation for 'mfr': Fisher Chemical L/0565/48

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 105-74-8
Molecular Formula C24H46O4
Molecular Weight (g/mol) 398.628
MDL Number 8964
InChI Key YIVJZNGAASQVEM-UHFFFAOYSA-N
Synonym lauroyl peroxide, dilauroyl peroxide, dodecanoyl peroxide, peroxide, bis 1-oxododecyl, laurydol, laurox, alperox c, dodecanoic peroxyanhydride, dyp-97f, peroxyde de lauroyle
PubChem CID 7773
ChEBI CHEBI:82479
IUPAC Name dodecanoyl dodecaneperoxoate
SMILES CCCCCCCCCCCC(=O)OOC(=O)CCCCCCCCCCC

Specifications

Melting Point 53°C
Color White
Packaging Plastic powder jar
Quantity 100 g
Formula Weight 398.63g/mol
Physical Form Powder
compliance-icons
Signal Word
  • Danger
Hazard Category
  • Organic peroxide Type C & D
Hazard Statement
  • H242-Heating may cause a fire.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P234-Keep only in original container.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P403-Store in a well-ventilated place.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.