missing translation for 'onlineSavingsMsg'
Learn More

Di-^m-chlorodichlorobis(ethylene)diplatinum(II), 97%, Thermo Scientific Chemicals

Product Code. 15496295
Click to view available options
Quantity:
250 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12073-36-8
Molecular Formula C4H8Cl4Pt2
Molecular Weight (g/mol) 588.08
MDL Number MFCD00058713
InChI Key XBLBDYCZFZESPN-UHFFFAOYSA-J
Synonym di-mu-chloro-dichlorobis ethylene diplatinum ii, zeise's dimer, ptcl2 c2h4 2, di-chloro-dichlorobis ethylene diplatinum ii, di-micro-chlorodichlorobis ethylene diplatinum ii
PubChem CID 11124780
SMILES [Cl-].[Cl-].[Cl-].[Cl-].[Pt++].[Pt++].C=C.C=C

Specifications

Quantity 250 mg
IUPAC Name dichloroplatinum;ethene
Formula Weight 588.1
Percent Purity 97%
Physical Form Crystalline
Chemical Name or Material Di-mu-chlorodichlorobis(ethylene)diplatinum(II)

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.