missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Di-n-butyl sulfoxide, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
Description
It is used as an active pharmaceutical intermediate and in the extraction of thorium(IV) from nitric acid solutions in xylene.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is used as an active pharmaceutical intermediate and in the extraction of thorium(IV) from nitric acid solutions in xylene.
Solubility
Slightly soluble in water.
Notes
Hygroscopic. Protect from water/moisture and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas. Protect from humidity and water.
It is used as an active pharmaceutical intermediate and in the extraction of thorium(IV) from nitric acid solutions in xylene.
Solubility
Slightly soluble in water.
Notes
Hygroscopic. Protect from water/moisture and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas. Protect from humidity and water.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Melting Point | 31°C to 34°C |
Density | 0.832 |
Boiling Point | 250°C |
Flash Point | 121°C (249°F) |
Refractive Index | 1.467 |
Quantity | 5 g |
Beilstein | 1747425 |
Sensitivity | Hygroscopic |
Solubility Information | Slightly soluble in water. |
Formula Weight | 162.3 |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'