Learn More
Di-n-butylamine, 99%, Thermo Scientific Chemicals
Di-n-butylamine, 99%, C8H19N, CAS Number-111-92-2, di-normal-butylamine, n-dibutylamine, n-butyl-1-butanamine, di-n-butyl amine, dibutilamina, dibutylamine, dibutyl amine, di-n-butylamine, dibutyl-amine, 1-butanamine, n-butyl, 2.5L, 98.5% min. (GC), 04, 157, 159.0 deg.C, Undesignated, 0.7600g/mL
£5.97 - £377.88
Chemical Identifiers
CAS | 111-92-2 |
---|---|
MDL Number | MFCD00009429 |
InChI Key | JQVDAXLFBXTEQA-UHFFFAOYSA-N |
Synonym | dibutylamine, di-n-butylamine, 1-butanamine, n-butyl, n-butyl-1-butanamine, dibutilamina, n-dibutylamine, di-n-butyl amine, dibutyl amine, di-normal-butylamine, dibutyl-amine |
PubChem CID | 8148 |
IUPAC Name | N-butylbutan-1-amine |
SMILES | CCCCNCCCC |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
11925171
|
Thermo Scientific Acros
112950050 |
5 mL | Glass bottle | N/A | |||||
10478993
|
Thermo Scientific Acros
112950500 |
50 mL | Glass bottle |
£29.35
50mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10617642
|
Thermo Scientific Acros
112950010 |
1 L | Glass Bottle |
£67.94
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10627642
|
Thermo Scientific Acros
112950025 |
2.5 L | Glass bottle |
£124.62
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11935171
|
Thermo Scientific Acros
112950100 |
10 L | Plastic drum |
£377.88
10 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
111-92-2 | |
JQVDAXLFBXTEQA-UHFFFAOYSA-N | |
8148 | |
CCCCNCCCC |
MFCD00009429 | |
dibutylamine, di-n-butylamine, 1-butanamine, n-butyl, n-butyl-1-butanamine, dibutilamina, n-dibutylamine, di-n-butyl amine, dibutyl amine, di-normal-butylamine, dibutyl-amine | |
N-butylbutan-1-amine |
Specifications
111-92-2 | |
Authentic | |
C8H19N | |
1.4160 to 1.4180 (20°C, 589nm) | |
04, 157 | |
15, 3037 | |
JQVDAXLFBXTEQA-UHFFFAOYSA-N | |
N-butylbutan-1-amine | |
8148 | |
≥98.5% | |
Di-n-butylamine |
0.7600g/mL | |
99% | |
[CH3(CH2)3]2NH | |
MFCD00009429 | |
01,209 | |
dibutylamine, di-n-butylamine, 1-butanamine, n-butyl, n-butyl-1-butanamine, dibutilamina, n-dibutylamine, di-n-butyl amine, dibutyl amine, di-normal-butylamine, dibutyl-amine | |
CCCCNCCCC | |
129.247 | |
129.24 | |
Liquid |
Safety and Handling
GHS H Statement:
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Toxic if swallowed.
Fatal if inhaled.
Flammable liquid and vapor.
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It i
GHS Signal Word: Danger
- Di-n-butylamine
Signal Word
- Danger
Hazard Category
- Flammable liquid Category 3
- Acute toxicity Category 3
- Acute toxicity Category 2
- Skin corrosion/irritation Category 1 B
- Serious eye damage/eye irritation Category 1
Hazard Statement
- H226-Flammable liquid and vapour.
- H301-Toxic if swallowed.
- H311-Toxic in contact with skin.
- H314-Causes severe skin burns and eye damage.
- H330-Fatal if inhaled.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only