missing translation for 'onlineSavingsMsg'
Learn More

Di-n-butylgermanium dichloride, 97%, Thermo Scientific Chemicals

Product Code. 11449006
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4593-81-1
Molecular Formula C8H18Cl2Ge
Molecular Weight (g/mol) 257.762
MDL Number MFCD00013590
InChI Key VKMPTHGEGBKPBQ-UHFFFAOYSA-N
Synonym di-n-butylgermanium dichloride, dibutylgermanium dichloride, dichlorodibutylgermane, germane, dibutyldichloro, di-n-butylgermanedichloride, di-n-butyldichlorogermane, dibutyl dichloro germane, wln: g-ge-g4&4
PubChem CID 20723
IUPAC Name dibutyl(dichloro)germane
SMILES CCCC[Ge](CCCC)(Cl)Cl

Specifications

Density 1.208
Boiling Point 241°C to 243°C
Flash Point 107°C (225°F)
Linear Formula [CH3(CH2)3]2GeCl2
Refractive Index 1.473
Quantity 5 g
UN Number UN2922
Sensitivity Moisture sensitive
Formula Weight 257.73
Percent Purity 97%
Chemical Name or Material Di-n-butylgermanium dichloride
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'