Learn More
Di-n-butyltin bis(2,4-pentanedionate), 95%, Thermo Scientific Chemicals
£21.41 - £191.96
Chemical Identifiers
CAS | 22673-19-4 |
---|---|
Molecular Formula | C18H32O4Sn |
MDL Number | MFCD00077994 |
Synonym | dibutyltin bis acetylacetonate, di-n-butylbis 2,4-pentanedionate tin, dibutyltin bis 2,4-pentanedionate, 3-penten-2-one, 4,4'-dibutylstannylene bis oxy bis-, 3z,3'z, 3z-4-dibutyl 2z-4-oxopent-2-en-2-yl oxy stannyl oxy pent-3-en-2-one, z-4-dibutyl-z-1-methyl-3-oxo-but-1-enoxy stannyl oxypent-3-en-2-one |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11426786
|
Thermo Scientific Alfa Aesar
B23921.18 |
50 g |
£21.41
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11436786
|
Thermo Scientific Alfa Aesar
B23921.30 |
250 g |
£64.92
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11416786
|
Thermo Scientific Alfa Aesar
B23921.0B |
1000 g |
£191.96
1000g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
22673-19-4 | |
C18H32O4Sn | |
MFCD00077994 | |
dibutyltin bis acetylacetonate, di-n-butylbis 2,4-pentanedionate tin, dibutyltin bis 2,4-pentanedionate, 3-penten-2-one, 4,4'-dibutylstannylene bis oxy bis-, 3z,3'z, 3z-4-dibutyl 2z-4-oxopent-2-en-2-yl oxy stannyl oxy pent-3-en-2-one, z-4-dibutyl-z-1-methyl-3-oxo-but-1-enoxy stannyl oxypent-3-en-2-one |
Specifications
22673-19-4 | |
150°C | |
Weak | |
MFCD00077994 | |
dibutyltin bis acetylacetonate, di-n-butylbis 2,4-pentanedionate tin, dibutyltin bis 2,4-pentanedionate, 3-penten-2-one, 4,4'-dibutylstannylene bis oxy bis-, 3z,3'z, 3z-4-dibutyl 2z-4-oxopent-2-en-2-yl oxy stannyl oxy pent-3-en-2-one, z-4-dibutyl-z-1-methyl-3-oxo-but-1-enoxy stannyl oxypent-3-en-2-one | |
95% |
26°C | |
40°C (104°F) | |
C18H32O4Sn | |
UN1992 | |
431.16 | |
Di-n-butyltin bis(2,4-pentanedionate) |
Safety and Handling
GHS H Statement
H226-H302-H312-H332-H315-H319-H335
Flammable liquid and vapour.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Warning
missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, TOXIC, N.O.S.
missing translation for 'tsca' : TSCA
RUO – Research Use Only