missing translation for 'onlineSavingsMsg'
Learn More

Di-n-butyltin bis(2-ethylhexanoate), Thermo Scientific Chemicals

£30.25 - £206.03

Chemical Identifiers

CAS 2781-10-4
MDL Number MFCD00015261
PubChem CID 76044
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11344819
View Documents Promotion Details
Thermo Scientific Alfa Aesar
071126.06
5 g
£30.25
5g
Please to purchase this item. Need a web account? Register with us today!
11324819
View Documents Promotion Details
Thermo Scientific Alfa Aesar
071126.14
25 g
£69.75
25g
Please to purchase this item. Need a web account? Register with us today!
11334819
View Documents Promotion Details
Thermo Scientific Alfa Aesar
071126.22
100 g
£206.03
100g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Di-n-butyltin bis(2-ethylhexanoate) is used in silanol condensation reactions for caulk and sealant applications and in the production of silicones. It is also used in one or two component systems which are compatible with aromatic solvents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

2781-10-4
MFCD00015261
76044

Specifications

2781-10-4
1.070 g/mL
Odorless
MFCD00015261
s-trityl-l-cysteine, +-s-trityl-l-cysteine, h-cys trt-oh, s-tritylcysteine, tritylcysteine, 3-tritylthio-l-alanine, 2r-2-amino-3-triphenylmethyl sulfanyl propanoic acid, l-alanine, 3-tritylthio, stlc, s-triphenylmethyl-l-cysteine
DLMYFMLKORXJPO-UHFFFAOYNA-N
(2R)-2-amino-3-tritylsulfanylpropanoic acid
76044
Lumps
57°C to 59°C
220°C (2 mmHg)
C22H21NO2S
UN3146
Soluble in mineral oil, acetone, xylene
C1=CC=C(C=C1)C(C2=CC=CC=C2)(C3=CC=CC=C3)SCC(C(=O)O)N
363.48
519.34
Di-n-butyltin bis (2-ethylhexanoate)
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H310-H361
Toxic if swallowed.
Fatal in contact with skin.
Suspected of damaging fertility or the unborn child.

P201-P202-P260-P264b-P270-P281-P301+P310-P308+P313-P330-P501c

H301-H340-H360-H372-H500

missing translation for 'dotInformation' : Hazard Class: 6.1; Packaging Group: III

missing translation for 'einecsNumber' : 220-481-2

missing translation for 'rtecsNumber' : WH6714500

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only