Learn More
Di-n-butyltin dilaurate, 95%, Thermo Scientific Chemicals
£21.71 - £98.59
Chemical Identifiers
CAS | 77-58-7 |
---|---|
Molecular Formula | C32H64O4Sn |
MDL Number | MFCD00008963 |
Synonym | Di-n-butyltin didodecanoate |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11386319
|
Thermo Scientific Alfa Aesar
071130.14 |
25 g |
£21.71
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11354819
|
Thermo Scientific Alfa Aesar
071130.22 |
100 g |
£34.47
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11364819
|
Thermo Scientific Alfa Aesar
071130.36 |
500 g |
£98.59
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Di-n-butyltin dilaurate is used as a catalyst for the production of polyurethanes as well as for the transesterification reactions. It is involved in the vulcanization of silicones and a stabilizer in polyvinyl chloride (PVC). It acts as a rust inhibitor for polyurethanes, polyols, silicones and as a fuel additive.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
77-58-7 | |
C32H64O4Sn | |
MFCD00008963 | |
Di-n-butyltin didodecanoate |
Specifications
77-58-7 | |
Yellow | |
>204°C (12 mmHg) | |
C32H64O4Sn | |
1.473 | |
UN2788 | |
Di-n-butyltin didodecanoate | |
631.55 | |
Liquid |
<10°C | |
1.066 g/mL | |
235°C (455°F) | |
[CH3(CH2)3]2Sn[O2C(CH2)10CH3]2 | |
MFCD00008963 | |
14,3038 | |
Miscible with benzene and ether; Imiscible with water and methanol. | |
95% | |
Di-n-butyltin dilaurate |
Safety and Handling
GHS H Statement
H301-H331-H360-H341-H314-H318-H317
Toxic if swallowed.
Toxic if inhaled.
May damage fertility or the unborn child.
Suspected of causing genetic defects.
Causes severe skin burns and eye damage.
Causes serious eye damage.
May cause an allergic skin reaction.
P201-P202-P260-P264b-P270-P271-P272-P280g-P281-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P333+P313-P363-P501c
H314-H317-H335-H336-H341-H360FD-H370-H372
missing translation for 'dotInformation' : Hazard Class: 8; Packaging Group: III
missing translation for 'einecsNumber' : 201-039-8
missing translation for 'rtecsNumber' : WH7000000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only