Learn More
Di-n-butyltin oxide, Thermo Scientific Chemicals
£22.50 - £137.00
Chemical Identifiers
CAS | 818-08-6 |
---|---|
Molecular Formula | C8H18OSn |
Molecular Weight (g/mol) | 248.941 |
MDL Number | MFCD00001992 |
InChI Key | JGFBRKRYDCGYKD-UHFFFAOYSA-N |
Synonym | dibutyltin oxide, stannane, dibutyloxo, dibutyl oxo tin, dibutyloxostannane, di-n-butyltin oxide, tin, dibutyloxo, dibutylstannane oxide, dibutyltinoxide, dibutyltin iv oxide, dibutyloxide of tin |
PubChem CID | 61221 |
IUPAC Name | dibutyl(oxo)tin |
SMILES | CCCC[Sn](=O)CCCC |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11387717
|
Thermo Scientific Alfa Aesar
|
50 g |
£22.50
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11397717
|
Thermo Scientific Alfa Aesar
|
250 g | N/A | ||||||
11377717
|
Thermo Scientific Alfa Aesar
|
1 kg |
£137.00
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Di-n-butyltin oxide is used as a reagent and a catalyst. It is particularly useful in regioselective alkylation, acylation and sulfonation reactions for starting materials containing alcohol functional groups. It is used in studies pertaining to discovering novel antitumor agents, antifungal activity.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
818-08-6 | |
248.941 | |
JGFBRKRYDCGYKD-UHFFFAOYSA-N | |
61221 | |
CCCC[Sn](=O)CCCC |
C8H18OSn | |
MFCD00001992 | |
dibutyltin oxide, stannane, dibutyloxo, dibutyl oxo tin, dibutyloxostannane, di-n-butyltin oxide, tin, dibutyloxo, dibutylstannane oxide, dibutyltinoxide, dibutyltin iv oxide, dibutyloxide of tin | |
dibutyl(oxo)tin |
Specifications
818-08-6 | |
White | |
Odorless | |
[CH3(CH2)3]2SnO | |
UN3146 | |
dibutyltin oxide, stannane, dibutyloxo, dibutyl oxo tin, dibutyloxostannane, di-n-butyltin oxide, tin, dibutyloxo, dibutylstannane oxide, dibutyltinoxide, dibutyltin iv oxide, dibutyloxide of tin | |
JGFBRKRYDCGYKD-UHFFFAOYSA-N | |
dibutyl(oxo)tin | |
61221 | |
Powder |
>300°C | |
1.58 g/mL | |
C8H18OSn | |
MFCD00001992 | |
4126243 | |
Soluble in water 4.0g/L @ 20°C. | |
CCCC[Sn](=O)CCCC | |
248.941 | |
248.92 | |
Di-n-butyltin oxide |
Safety and Handling
GHS H Statement
H300-H319
Fatal if swallowed.
Causes serious eye irritation.
P201-P202-P260-P264b-P270-P272-P280g-P281-P301+P310-P302+P352-P305+P351+P338-P307+P311-P310-P330-P333+P313-P362-P501c
H301-H315-H317-H318-H341-H360-H370-H372
missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: ORGANOTIN COMPOUNDS, SOLID, N.O.S.
missing translation for 'einecsNumber' : 212-449-1
missing translation for 'rtecsNumber' : WH7175000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only