missing translation for 'onlineSavingsMsg'
Learn More
Di-n-butyltin oxide, 98%, Thermo Scientific Chemicals Product Code.: 11383546

Di-n-butyltin oxide, 98%, Thermo Scientific Chemicals

Product Code. 11383546
500 g
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy

Product Code. 11383546

Brand: Thermo Scientific Alfa Aesar L14491.36

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 818-08-6
Molecular Formula C8H18OSn
Molecular Weight (g/mol) 248.941
MDL Number MFCD00001992
InChI Key JGFBRKRYDCGYKD-UHFFFAOYSA-N
Synonym dibutyltin oxide, stannane, dibutyloxo, dibutyl oxo tin, dibutyloxostannane, di-n-butyltin oxide, tin, dibutyloxo, dibutylstannane oxide, dibutyltinoxide, dibutyltin iv oxide, dibutyloxide of tin
PubChem CID 61221
IUPAC Name dibutyl(oxo)tin
SMILES CCCC[Sn](=O)CCCC

Specifications

Melting Point >300°C
Density 1.58
Odor Odorless
Linear Formula [CH3(CH2)3]2SnO
Quantity 500 g
UN Number UN3146
Beilstein 4126243
Formula Weight 248.94
Percent Purity 98%
Chemical Name or Material Di-n-butyltin oxide

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberR06F029 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberL14491.14,L14491.36,L14491.22
Lot NumberUSLF004913 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberL14491.14,L14491.36,L14491.22
Lot Number10150532 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog NumberL14491.14,L14491.36,L14491.22
Lot NumberH21U009 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog NumberL14491.14,L14491.36,L14491.22
Lot NumberK12W035 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog NumberL14491.14,L14491.36,L14491.22

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Di-n-butyltin oxide, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.