missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Di-n-octyl phthalate, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
25 mL
100 mL
500 mL
Description
Di-n-octyl phthalate is used as a plasticizer for many resins and elastomers. It acts as an additive. It is also used for medical tubing and blood storage bags, wire and cables, carpet back coating, floor tile and in cosmetics.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Density | 0.98g/mL |
Assay Percent Range | 98% |
Quantity | 25 mL |
Merck Index | 14,2864 |
Formula Weight | 390.56 |
Percent Purity | 98% |
Physical Form | Liquid |
Chemical Name or Material | Di-n-octyl phthalate |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'