missing translation for 'onlineSavingsMsg'
Learn More

Di-n-octyltin oxide, Thermo Scientific™

Product Code. 10314402
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 870-08-6
Molecular Formula C16H34OSn
Molecular Weight (g/mol) 361.16
MDL Number MFCD00013839
InChI Key LQRUPWUPINJLMU-UHFFFAOYSA-N
Synonym di-n-octyltin oxide, stannane, dioctyloxo, dioctyltin oxide, dioctyloxostannane, tin, dioctyloxo, dioctyl oxo stannane, stannane, oxodioctyl, di-n-octyl-zinn oxyd, tin, dioctyl-, oxide, dioctyl oxo tin
PubChem CID 93563
IUPAC Name dioctyl(oxo)tin
SMILES CCCCCCCC[Sn](=O)CCCCCCCC

Specifications

Color White
Infrared Spectrum Authentic
Packaging Glass Bottle
Quantity 25 g
Solubility Information Solubility in water: insoluble in water
Formula Weight 361.14
Percent Purity ≥32.0% (Sn)
Physical Form Crystalline Powder or Granules
Chemical Name or Material Di-n-octyltin oxide
compliance-icons
Product Identifier
  • Di-n-octyltin oxide
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'